Learn More
ACROBiosystems ACROBiosystems Cynomolgus Nectin-4 Protein, His Tag
Catalog No. 501956558
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus Nectin-4, His Tag (NE4-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # XP_005541277.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.0 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6558 Supplier ACROBiosystems Supplier No. NE4C52H41MG
Recombinant Protein;1MG;Cynomolgus Nectin-4, His Tag (NE4-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # XP_005541277.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.0 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.