Learn More
Genscript Corporation Nectin-4 HisCynomolgus
Catalog No. 5000057238
Encompass
Nectin-4 seems to be involved in cell adhesion through trans-homophilic and -heterophilic interactions the latter including specifically interactions with NECTIN1 Does not act as receptor for alpha-herpesvirus entry into cells (Microbial infection) Acts as a receptor for measles virus
Catalog No. 50-000-57238 Supplier Genscript Corporation Supplier No. Z03827500
Nectin-4 seems to be involved in cell adhesion through trans-homophilic and -heterophilic interactions the latter including specifically interactions with NECTIN1 Does not act as receptor for alpha-herpesvirus entry into cells (Microbial infection) Acts as a receptor for measles virus