Learn More
ACROBiosystems Recombinant Protein;Human Nectin-1 / PVRL1 / CD111 Protein;HEK293;100UG;PV1,PVRL1,Nectin-1,CD111,CLPED1,ED4,HIgR,HVEC,OFC7,PRR,PRR1,PVRR,PVRR1,SK-12,PVRL-1,Nectin 1
Supplier: ACROBiosystems PV1H5223100UG
Recombinant Protein;Human Nectin-1 / PVRL1 / CD111 Protein;100UG;Human Nectin-1, His Tag (PV1-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 334 (Accession # NP_002846).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.8 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.