Learn More
ACROBiosystems Recombinant Protein;Human Nectin-2 / CD112 Protein, Mouse IgG2a Fc Tag;HEK293;100UG;CD2,CD112,Nectin-2,PVRL2,PVRL,Nectin
Catalog No. 501657464
Encompass_Preferred
Recombinant Protein;Human Nectin-2 / CD112 Protein, Mouse IgG2a Fc Tag;100UG;Human Nectin-2, Mouse IgG2a Fc Tag (CD2-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 62.4 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human DNAM-1, Fc Tag (Cat. No. DN1-H5257) at 5 μg/mL (100 μL/well) can bind Human Nectin-2, Mouse IgG2a Fc Tag (Cat. No. CD2-H5257) with a linear range of 2-20 ng/mL (QC tested).
Catalog No. 50-165-7464 Supplier ACROBiosystems Supplier No. CD2H5257100UG
Recombinant Protein;Human Nectin-2 / CD112 Protein, Mouse IgG2a Fc Tag;100UG;Human Nectin-2, Mouse IgG2a Fc Tag (CD2-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 62.4 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human DNAM-1, Fc Tag (Cat. No. DN1-H5257) at 5 μg/mL (100 μL/well) can bind Human Nectin-2, Mouse IgG2a Fc Tag (Cat. No. CD2-H5257) with a linear range of 2-20 ng/mL (QC tested).