Learn More
Biomatik Corporation Nectin4 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11459
Encompass_Preferred
Nectin4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NECTIN4. The Nectin4 Polyclonal Antibody has been validated for the following applications: ELISA, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nectin-4 Protein (91-199Aa). Applications: ELISA, IF. Target Synonyms: NECTIN4. Accession Number: Q96NY8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5207