Learn More
Empire Genomics NECTIN4/CON1 FISH Probe, 20 tests, Orange/Green labeling, 40 μL
Catalog No. NC3456086
Encompass
The EMPIRE GENOMICS NECTIN4/CON1 FISH Probe is a specialized Fluorescence In Situ Hybridization (FISH) probe designed for the detection of NECTIN4 gene copy number variations. This essential tool incorporates an internal control (CON1) to ensure accurate and reliable analysis of chromosomal abnormalities. The probe features distinct Orange and Green fluorophores, enabling clear differential detection for enhanced visibility during studies of genetic variations.
  • Target gene is NECTIN4.
  • Designed for detecting copy number variations.
  • Includes an internal control probe (CON1).
  • Type of probe is FISH.
  • Labeling features Orange and Green fluorophores.
  • Quantity provided is 20 tests (40 μL).
  • Orange fluorophore has excitation maximum of 552 nm and emission maximum of 576 nm.
  • Green fluorophore has excitation maximum of 491 nm and emission maximum of 516 nm.
Catalog No. NC3456086 Supplier Empire Genomics Supplier No. NECTIN4CHR0120OGR
The EMPIRE GENOMICS NECTIN4/CON1 FISH Probe is a specialized Fluorescence In Situ Hybridization (FISH) probe designed for the detection of NECTIN4 gene copy number variations. This essential tool incorporates an internal control (CON1) to ensure accurate and reliable analysis of chromosomal abnormalities. The probe features distinct Orange and Green fluorophores, enabling clear differential detection for enhanced visibility during studies of genetic variations.
  • Target gene is NECTIN4.
  • Designed for detecting copy number variations.
  • Includes an internal control probe (CON1).
  • Type of probe is FISH.
  • Labeling features Orange and Green fluorophores.
  • Quantity provided is 20 tests (40 μL).
  • Orange fluorophore has excitation maximum of 552 nm and emission maximum of 576 nm.
  • Green fluorophore has excitation maximum of 491 nm and emission maximum of 516 nm.