Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Empire Genomics NECTIN4 vB FISH Probe, Detects NECTIN4 gene copy number variations, 20 tests per kit, Orange-dUTP, Green-dUTP

Catalog No. NC3453863
Encompass

Empire Genomics' NECTIN4/CON1 vB FISH Probe is a specialized tool designed for detecting NECTIN4 gene copy number variations in scientific research. This probe incorporates an internal control to enhance accuracy and reliability while being pre-labeled with distinct Orange and Green fluorophores for clear signal differentiation. It is supplied as a concentrate, offering 20 tests per kit.

  • Target gene: NECTIN4
  • Application: Copy number variation detection
  • Internal control: Yes
  • Fluorescent labels: Orange-dUTP, Green-dUTP
  • Orange label excitation: 552 nm
  • Orange label emission: 576 nm
  • Green label excitation: 491 nm
  • Green label emission: 516 nm
  • Tests per kit: 20 tests
  • Probe volume per test: 2 μL
  • Product type: FISH probe
  • Regulatory status: Research Use Only (RUO)

Catalog No. NC3453863 Supplier Empire Genomics Supplier No. NECTIN4VBCHR0120OGR
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Empire Genomics' NECTIN4/CON1 vB FISH Probe is a specialized tool designed for detecting NECTIN4 gene copy number variations in scientific research. This probe incorporates an internal control to enhance accuracy and reliability while being pre-labeled with distinct Orange and Green fluorophores for clear signal differentiation. It is supplied as a concentrate, offering 20 tests per kit.

  • Target gene: NECTIN4
  • Application: Copy number variation detection
  • Internal control: Yes
  • Fluorescent labels: Orange-dUTP, Green-dUTP
  • Orange label excitation: 552 nm
  • Orange label emission: 576 nm
  • Green label excitation: 491 nm
  • Green label emission: 516 nm
  • Tests per kit: 20 tests
  • Probe volume per test: 2 μL
  • Product type: FISH probe
  • Regulatory status: Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.