Learn More
Sino Biological Recombinant Human Nectin-4 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 19771H08H50
Encompass_Preferred
A DNA sequence encoding the human NECTIN4 (NP_112178.2) (Met1-Ser349) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5129