Learn More
Aviva Systems Biology Corp NEDD4L Antibody - middle region (ARP43192_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43192_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NEDD4L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NEDD4L. Alternate names: FLJ33870, KIAA0439, RSP5, hNedd4-2, NEDD4-2, NEDD4.2, hNEDD4-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58459