Learn More
Enzo Life Sciences NEDD8-AMC (25 µg)
SDP
Catalog No. 502012227
Encompass_Preferred
NEDD8-AMC is a fluorogenic substrate for deNEDDylating enzymes, including NEDP1 and UCH-L3. It is a particularly useful reagent for the study of deNEDDylating activity where detection sensitivity or continuous monitoring of activity is essential. NEDD8-AMC is a fluorogenic substrate for deNEDDylating enzymes, including NEDP1 and UCH-L3. It is a particularly useful reagent for the study of deNEDDylating activity where detection sensitivity or continuous monitoring of activity is essential. Concentration: 1 mg/ml. Purity: ≥95% (HPLC). Formulation: Solution in DMSO. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2227 Supplier Enzo Life Sciences Supplier No. BMLUW00500025
NEDD8-AMC is a fluorogenic substrate for deNEDDylating enzymes, including NEDP1 and UCH-L3. It is a particularly useful reagent for the study of deNEDDylating activity where detection sensitivity or continuous monitoring of activity is essential. NEDD8-AMC is a fluorogenic substrate for deNEDDylating enzymes, including NEDP1 and UCH-L3. It is a particularly useful reagent for the study of deNEDDylating activity where detection sensitivity or continuous monitoring of activity is essential. Concentration: 1 mg/ml. Purity: ≥95% (HPLC). Formulation: Solution in DMSO. Source: Produced in E. coli. Long Term Storage: -80°C.