Learn More
Selleck Chemical LLC NEDD8 inhibitor M22 S3400-25mg
SDP
Catalog No. 503109315
Encompass_Preferred
NEDD8 inhibitor M22 (NAE-IN-M22) is a novel selective NEDD8 activating enzyme (NAE) inhibitor with GI50 of 5 5 M in A549 cells
Catalog No. 50-310-9315 Supplier Selleck Chemical LLC Supplier No. S340025MG
Add to Cart
Add to Cart
NEDD8 inhibitor M22 (NAE-IN-M22) is a novel selective NEDD8 activating enzyme (NAE) inhibitor with GI50 of 5 5 M in A549 cells