Learn More
Enzo Life Sciences NEDD8 polyclonal antibody (25µl)
SDP
Supplier:  Enzo Life Sciences ENZABS3760025
Encompass_Preferred
Isotype: IgG. Immunogen: Recombinant human NEDD8. Species reactivity: Human, Mouse, Rat. Applications: IHC, WB. Purity: Affinity-purified. Formulation: Liquid. In PBS, pH 7.3, containing 0.05% sodium azide and 50% glycerol.
Catalog No. 50-201-2903