Learn More
Aviva Systems Biology Corp NEDD9 Antibody - middle region (ARP51211_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51211_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NEDD9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NEDD9. Alternate names: CAS-L, CASL, HEF1, dJ49G10.2, dJ761I2.1, CAS2, CASS2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60517