Learn More
Metrohm USA NEEDLE
SDP
Supplier:  Metrohm USA 62816030
Encompass
Needle for filling in mercury into the Multi-Mode Electrode and for sample addition in Karl Fischer titration.
Catalog No. NC0329657
 Restricted Item