Learn More
Waters Corp Waters Needle, 15 μL, Stainless Steel, with Guide, 18,000 psi
Catalog No. 502039627 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement needle assembly features a 15 μL stainless steel needle with a guide and seat, rated for 18,000 psi. It is designed for use in ACQUITY I-Class Flow Through Needle (FTN) Sample Managers.
  • Replacement needle assembly
  • 15 μL capacity
  • Stainless steel construction
  • Includes guide and seat
  • Rated for 18,000 psi
  • Compatible with flow through needle sample managers
  • Designed for UPLC systems
Catalog No. 50-203-9627 Supplier Waters Corp Supplier No. 700008977
This replacement needle assembly features a 15 μL stainless steel needle with a guide and seat, rated for 18,000 psi. It is designed for use in ACQUITY I-Class Flow Through Needle (FTN) Sample Managers.
  • Replacement needle assembly
  • 15 μL capacity
  • Stainless steel construction
  • Includes guide and seat
  • Rated for 18,000 psi
  • Compatible with flow through needle sample managers
  • Designed for UPLC systems