Learn More
Waters Corp Waters Corporation Needle, Stainless Steel, 15μL
Catalog No. 50791671 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This optional stainless steel needle is designed for use with the ACQUITY UPLC Sample Manager.
  • Made from stainless steel
  • Features a 15μL volume
  • Designed for UPLC system applications
  • Compatible with premier system architectures
  • Functions as a component for column managers
Catalog No. 50-791-671 Supplier Waters Corp Supplier No. 205000363
This optional stainless steel needle is designed for use with the ACQUITY UPLC Sample Manager.
  • Made from stainless steel
  • Features a 15μL volume
  • Designed for UPLC system applications
  • Compatible with premier system architectures
  • Functions as a component for column managers