Learn More
Med Vet International Terumo Needle, 18G x 1.5in, Hypodermic, 100/Box, NN1838R
Catalog No. 502522578
Encompass_Preferred
This Terumo NN1838R Needle comes with a 18G X 1 1/2' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-252-2578 Supplier Med Vet International Supplier No. 1838RN
This Terumo NN1838R Needle comes with a 18G X 1 1/2' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX