Learn More
Med Vet International Nipro Needle, 19G x 1in, Hypodermic, 100/Box, AH+1925
Catalog No. 502522467
Encompass_Preferred
This Nipro AH+1925 Needle is a 19G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-252-2467 Supplier Med Vet International Supplier No. AH1925
This Nipro AH+1925 Needle is a 19G X 1' Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles allow rapid and efficient drug delivery. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX