Learn More
Med Vet International BD Needle, 23G X 1in, Hypodermic, 100/BX, 305145
Catalog No. 503043507
Encompass_Preferred
This BD 305145 Needle is a 23G X 1" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX
Catalog No. 50-304-3507 Supplier Med Vet International Supplier No. 305145
This BD 305145 Needle is a 23G X 1" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 100/BX