Learn More
Med Vet International BD Hypodermic Needles, 25G x 1-in., 100/Box
Catalog No. 503044994
Encompass_Preferred
These needles are regular wall. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-304-4994 Supplier Med Vet International Supplier No. 305125
These needles are regular wall. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.