Learn More
Med Vet International BD SafetyGlide™ Needle, 25G x 1" , 50/Box
Catalog No. 503043647
Encompass_Preferred
This BD 305916 Needle is a 25G X 1" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 50/BX
Catalog No. 50-304-3647 Supplier Med Vet International Supplier No. 305916
This BD 305916 Needle is a 25G X 1" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 50/BX