Learn More
Braintree Scientific Blunt Tip Needle Non-Sterile, 27ga - 1" w/std hub, 12/pk
SDP
Catalog No. NC9250452
Encompass
Blunt Tip Needle, 27 Gauge 1 in. Stainless Steel Hypodermic Needle with Standard Plastic Luer Hub, used to Connect Syringe Directly to Catheter Port for Intravascular Substance Administration, Blood Sampling and Catheter Flushing **Sold for research purposes only and not for human use**
Catalog No. NC9250452 Supplier Braintree Scientific Supplier No. NB27G
Blunt Tip Needle, 27 Gauge 1 in. Stainless Steel Hypodermic Needle with Standard Plastic Luer Hub, used to Connect Syringe Directly to Catheter Port for Intravascular Substance Administration, Blood Sampling and Catheter Flushing **Sold for research purposes only and not for human use**