Learn More
Jensen Global Inc NEEDLE 27G X 1/2" CLEAR 50/PAC
Catalog No. NC0897031
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
NT Series premium dispensing tips are manufactured using the highest quality standards and are angled at 45 or 90 degrees. The bend is made at 3/16" (4.763 mm) from the tip for 0.5", at 1/4" (6.350 mm) for 1.0", and at 5/16" (7.948 mm) for 1.5" long tips. The polished cannula and burr-free design provide the user accuracy and durability. With a polypropylene hub and 304 stainless steel cannula, our NT series tips provide the smoothest flow for precise dispensing applications. Available in boxes of 50 or bags of 1000. (non sterile / non medical) Gauge: 27 Color: Clear ID: 0.008" (0.210 mm) OD: 0.016" (0.410 mm)
Catalog No. NC0897031 Supplier Jensen Global Inc Supplier No. JG270.5HPX
This item has a minimum qty of 3 per supplier requirements.
NT Series premium dispensing tips are manufactured using the highest quality standards and are angled at 45 or 90 degrees. The bend is made at 3/16" (4.763 mm) from the tip for 0.5", at 1/4" (6.350 mm) for 1.0", and at 5/16" (7.948 mm) for 1.5" long tips. The polished cannula and burr-free design provide the user accuracy and durability. With a polypropylene hub and 304 stainless steel cannula, our NT series tips provide the smoothest flow for precise dispensing applications. Available in boxes of 50 or bags of 1000. (non sterile / non medical) Gauge: 27 Color: Clear ID: 0.008" (0.210 mm) OD: 0.016" (0.410 mm)