Learn More
Waters Corp Needle Assembly, 250μm, PEEKsil, UPLC System
Catalog No. 502392024 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement needle assembly features a 250μm PEEKsil construction. It is an optional needle assembly designed for use with the ACQUITY Flow Through Needle (FTN) Sample Manager and UPLC systems.
  • Designed for UPLC systems
  • Features 250μm PEEKsil material
  • Compatible with ACQUITY Flow Through Needle (FTN) Sample Manager
  • Available as a spare part kit
  • Supplied as 1 unit per package
  • Recommended for annual replacement
Catalog No. 50-239-2024 Supplier Waters Corp Supplier No. 700005179
This replacement needle assembly features a 250μm PEEKsil construction. It is an optional needle assembly designed for use with the ACQUITY Flow Through Needle (FTN) Sample Manager and UPLC systems.
  • Designed for UPLC systems
  • Features 250μm PEEKsil material
  • Compatible with ACQUITY Flow Through Needle (FTN) Sample Manager
  • Available as a spare part kit
  • Supplied as 1 unit per package
  • Recommended for annual replacement