Learn More
Waters Corp Waters Needle Assembly, 30 μL, Stainless Steel, 0.010 in. ID
Catalog No. NC0789441 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement 30μL stainless steel sample needle assembly includes a guide and is an optional component for the ACQUITY Flow Through Needle (FTN) Sample Manager. It features a 0.010 in. ID and is designed for use with UPLC and HPLC applications to reduce cycle times for larger extension loops.
  • 30μL capacity and stainless steel construction
  • Includes a guide
  • Optional assembly for flow through needle (FTN) sample manager
  • Inner diameter of 0.010 in.
  • Suitable for UPLC and HPLC applications to reduce cycle times for larger extension loops
  • Compatible with various HPLC and UPLC systems and sample managers
Catalog No. NC0789441 Supplier Waters Corp Supplier No. 700005279
This replacement 30μL stainless steel sample needle assembly includes a guide and is an optional component for the ACQUITY Flow Through Needle (FTN) Sample Manager. It features a 0.010 in. ID and is designed for use with UPLC and HPLC applications to reduce cycle times for larger extension loops.
  • 30μL capacity and stainless steel construction
  • Includes a guide
  • Optional assembly for flow through needle (FTN) sample manager
  • Inner diameter of 0.010 in.
  • Suitable for UPLC and HPLC applications to reduce cycle times for larger extension loops
  • Compatible with various HPLC and UPLC systems and sample managers