Learn More
Waters Corp Needle Assembly, MP35N, 0.007 ID
Supplier: Waters Corp 700009406
This replacement needle assembly is made of MP35N with a 0.007 ID. It is designed as a standard fraction collector needle for a Fraction Manager Analytical.
- Made of MP35N material.
- Features a 0.007 ID.
- Functions as a standard fraction collector needle.
- Suitable for Fraction Manager Analytical.
- Designed for UPLC systems.
- Recommended for annual replacement.