Learn More
Waters Corp Waters Needle Assembly, MP35N, 0.010 in. ID
Catalog No. 502039793 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement needle assembly, constructed from MP35N, features a 0.010 in. ID. It functions as an optional fraction collector needle in a Fraction Manager Analytical, particularly when higher flow rates are required. This needle must be used in conjunction with part number 700010345.
  • Serves as an optional fraction collector needle
  • Designed for use in a Fraction Manager Analytical when higher flow rates are needed
Catalog No. 50-203-9793 Supplier Waters Corp Supplier No. 700010339
This replacement needle assembly, constructed from MP35N, features a 0.010 in. ID. It functions as an optional fraction collector needle in a Fraction Manager Analytical, particularly when higher flow rates are required. This needle must be used in conjunction with part number 700010345.
  • Serves as an optional fraction collector needle
  • Designed for use in a Fraction Manager Analytical when higher flow rates are needed