Learn More
Sciencix PEEK Needle Assembly, High Quality Replacement for Waters, 30 µL Volume
SDP
Catalog No. NC3590222
Encompass_Preferred
The PEEK Needle Assembly is a high-quality replacement part designed for Waters ACQUITY and ACQUITY UPLC Sample Managers, ensuring precise sample handling. Constructed with durable PEEK and stainless steel components, it guarantees reliable performance in scientific applications. With a volume of 30 µL, it is a cost-effective option that seamlessly integrates with existing systems, making it an ideal choice for labs seeking efficiency and precision in their analytical processes.
Catalog No. NC3590222 Supplier Sciencix Supplier No. CTSA11151
The PEEK Needle Assembly is a high-quality replacement part designed for Waters ACQUITY and ACQUITY UPLC Sample Managers, ensuring precise sample handling. Constructed with durable PEEK and stainless steel components, it guarantees reliable performance in scientific applications. With a volume of 30 µL, it is a cost-effective option that seamlessly integrates with existing systems, making it an ideal choice for labs seeking efficiency and precision in their analytical processes.