Learn More
World Precision Instrument Needle Blades
SDP
Supplier:  World Precision Instrument 501249
Encompass
These Miniature Scalpel Blades and Handles (sold separately) can be combined for use as small scalpels which are handy in a life science laboratory for microdissection or small animal surgeries. We offer both a knurled, round handle and a hex handle. Both give you an excellent grip of the scalpel. These handles are 10 cm (3.9") long. We also offer 3 styles of blades. Needle blades are 3 cm long and come in packages of 6. The miniature edge blades have a small curved blade. They are 3.5 cm long and come in packages of 12. And we have a miniature edge blade with a round tip. They are also 3.5 cm long and are sold in packages of 12. The handles and blades are all made of stainless steel. The blades come sterile and are disposable. The handles may be reused.
Catalog No. 50-822-605