Learn More
Trajan Scientific Needle Cone Medical Lab 2/PK
SDP
Catalog No. 5000494185
Encompass_Preferred
5 cm 0 47 mm OD cone tipped needle for 10 L syringes PK2
Catalog No. 50-004-94185 Supplier Trajan Scientific Supplier No. 037010
5 cm 0 47 mm OD cone tipped needle for 10 L syringes PK2