Learn More
Trajan Scientific Needle Cone Medical Lab 2/PK
SDP
Catalog No. 5000494768
Encompass_Preferred
5 cm 0 63 mm OD cone tipped needle for 10 L syringes PK2
Catalog No. 50-004-94768 Supplier Trajan Scientific Supplier No. 037011
Add to Cart
Add to Cart
5 cm 0 63 mm OD cone tipped needle for 10 L syringes PK2