Learn More
Trajan Scientific Needle Cone Medical Lab 3/PK
SDP
Catalog No. 5000494852
Encompass_Preferred
8 cm 0 47 mm OD cone tipped needle for 10 L CTC Analytics and Thermo Scientific syringes PK3
Catalog No. 50-004-94852 Supplier Trajan Scientific Supplier No. 031535
8 cm 0 47 mm OD cone tipped needle for 10 L CTC Analytics and Thermo Scientific syringes PK3