Learn More
Trajan Scientific Needle Diamond MS Bevel 1/EA
SDP
Supplier:  Trajan Scientific 0355323
Encompass_Preferred
Ultra smooth hydrophobic coated 50 mm 0 63 mm OD 0 32 mm ID bevel tipped needle for 100-500 L manual MS syringes
Catalog No. 50-004-94970