Learn More
Trajan Scientific Needle Diamond MS Bevel 1/EA
Supplier: Trajan Scientific 0355321
Ultra smooth hydrophobic coated 50 mm 0 50 mm OD 0 20 mm ID bevel tipped needle for 10-50 L manual MS syringes