Learn More
Trajan Scientific Needle Diamond MS Bevel 1/EA
SDP
Catalog No. 5000494738
Encompass_Preferred
Ultra smooth 50 mm 0 47 mm OD 0 11 mm ID bevel tipped needle for 10-50 L manual MS syringes
Catalog No. 50-004-94738 Supplier Trajan Scientific Supplier No. 0355330
Ultra smooth 50 mm 0 47 mm OD 0 11 mm ID bevel tipped needle for 10-50 L manual MS syringes