Learn More
Trajan Scientific Needle Diamond MS LC 1/EA
SDP
Catalog No. 5000494803
Encompass_Preferred
Ultra smooth 57 mm 0 72 mm OD 0 17 mm ID LC tipped needle for 10-50 L manual and autosampler MS syringes
Catalog No. 50-004-94803 Supplier Trajan Scientific Supplier No. 0355235
Ultra smooth 57 mm 0 72 mm OD 0 17 mm ID LC tipped needle for 10-50 L manual and autosampler MS syringes