Learn More
Trajan Scientific Needle Diamond MS LC 1/EA
SDP
Supplier:  Trajan Scientific 0355236
Encompass_Preferred
Ultra smooth 57 mm 0 72 mm OD 0 17 mm ID LC tipped needle for 100-500 L manual and autosampler MS syringes
Catalog No. 50-004-94841