Learn More
Trajan Scientific Needle Diamond MS LC 1/EA
SDP
Supplier:  Trajan Scientific 0355215
Encompass_Preferred
Ultra smooth hydrophilic coated 57 mm 0 72 mm OD 0 17 mm ID LC tipped needle for 10-50 L manual and autosampler MS syringes
Catalog No. 50-004-94756