Learn More
Trajan Scientific Needle Diamond MS LC 1/EA
SDP
Catalog No. 5000494810
Encompass_Preferred
Ultra smooth hydrophobic coated 51 mm 0 72 mm OD 0 37 mm ID LC tipped needle for 100-500 L manual and autosampler MS syringes
Catalog No. 50-004-94810 Supplier Trajan Scientific Supplier No. 0355223
Ultra smooth hydrophobic coated 51 mm 0 72 mm OD 0 37 mm ID LC tipped needle for 100-500 L manual and autosampler MS syringes