Learn More
Trajan Scientific Needle Diamond MS LC 1/EA
Supplier: Trajan Scientific 0355211
Ultra smooth hydrophilic coated 51 mm 0 72 mm OD 0 17 mm ID LC tipped needle for 100-500 L manual and autosampler MS syringes