Learn More
Trajan Scientific Needle Diamond MS LC 1/EA
SDP
Catalog No. 5000494745
Encompass_Preferred
Ultra smooth hydrophilic coated 57 mm 0 72 mm OD 0 17 mm ID LC tipped needle for 100-500 L manual and autosampler MS syringes
Catalog No. 50-004-94745 Supplier Trajan Scientific Supplier No. 0355216
Ultra smooth hydrophilic coated 57 mm 0 72 mm OD 0 17 mm ID LC tipped needle for 100-500 L manual and autosampler MS syringes