Learn More
Trajan Scientific Needle Diamond MS Medical 1/EA
SDP
Catalog No. 5000494874
Encompass_Preferred
Ultra smooth hydrophilic coated 42 mm 0 63 mm OD 0 11 mm ID cone tipped needle for 10-50 L Agilent and Shimadzu GC autosampler MS syringes
Catalog No. 50-004-94874 Supplier Trajan Scientific Supplier No. 0355411
Ultra smooth hydrophilic coated 42 mm 0 63 mm OD 0 11 mm ID cone tipped needle for 10-50 L Agilent and Shimadzu GC autosampler MS syringes