Learn More
Trajan Scientific Needle Diamond MS Medical 1/EA
SDP
Supplier:  Trajan Scientific 0355612
Encompass_Preferred
Ultra smooth hydrophilic coated 57 mm 0 63 mm OD 0 11 mm ID cone tipped needle for 10-50 L CTC Analytics PAL RTC (PAL3) Shimadzu AOC-6000 Thermo Scientific TriPlus RSH and Agilent PAL3 syringe GC autosampler MS syringes
Catalog No. 50-004-94854