Learn More
Trajan Scientific Needle Diamond MS Medical 1/EA
SDP
Catalog No. 5000494746
Encompass_Preferred
Ultra smooth 50 mm 0 63 mm OD 0 24 mm ID cone tipped needle for 100-500 L CTC GC autosampler MS syringes
Catalog No. 50-004-94746 Supplier Trajan Scientific Supplier No. 0355531
Ultra smooth 50 mm 0 63 mm OD 0 24 mm ID cone tipped needle for 100-500 L CTC GC autosampler MS syringes