Learn More
Trajan Scientific Needle Diamond MS Medical 1/EA
SDP
Supplier:  Trajan Scientific 0355521
Encompass_Preferred
Ultra smooth hydrophobic coated 50 mm 0 63 mm OD 0 24 mm ID cone tipped needle for 100-500 L CTC GC autosampler MS syringes
Catalog No. 50-004-94728