Learn More
Trajan Scientific Needle eVol Medical Lab 2/PK
SDP
Catalog No. 5000494751
Encompass_Preferred
5 cm 0 63 mm OD side hole dome tipped needle for 1-2 5 mL syringes (and 500 L eVol syringes) PK2
Catalog No. 50-004-94751 Supplier Trajan Scientific Supplier No. 039120
5 cm 0 63 mm OD side hole dome tipped needle for 1-2 5 mL syringes (and 500 L eVol syringes) PK2