Learn More
Trajan Scientific Needle eVol Medical Lab 5/PK
SDP
Catalog No. 5000493927
Encompass_Preferred
11 5 cm 0 63 mm OD bevel tipped needle for 1-2 5 mL syringes (and 500 L eVol syringes) PK5
Catalog No. 50-004-93927 Supplier Trajan Scientific Supplier No. 039160
11 5 cm 0 63 mm OD bevel tipped needle for 1-2 5 mL syringes (and 500 L eVol syringes) PK5