Learn More
Trajan Scientific Needle eVol Medical Lab 5/PK
SDP
Catalog No. 5000494498
Encompass_Preferred
5 cm 0 63 mm OD cone tipped needle for 5 L eVol syringes PK5
Catalog No. 50-004-94498 Supplier Trajan Scientific Supplier No. 036914
5 cm 0 63 mm OD cone tipped needle for 5 L eVol syringes PK5