Learn More
Trajan Scientific Needle eVol Medical Lab 5/PK
SDP
Catalog No. 5000494653
Encompass_Preferred
7 cm 0 50 mm OD bevel tipped needle for 5 L eVol syringes PK5
Catalog No. 50-004-94653 Supplier Trajan Scientific Supplier No. 036916
7 cm 0 50 mm OD bevel tipped needle for 5 L eVol syringes PK5