Learn More
Trajan Scientific Needle eVol Medical Lab 5/PK
SDP
Catalog No. 5000493825
Encompass_Preferred
5 1 cm 0 72 mm OD LC tipped needle for 5 L eVol syringes PK5
Catalog No. 50-004-93825 Supplier Trajan Scientific Supplier No. 036912
5 1 cm 0 72 mm OD LC tipped needle for 5 L eVol syringes PK5